Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001569

Sigma-Aldrich

Glycerol

USP, 99.0-101.0%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein/REAXYS Number:
635685
EC Number:
MDL number:
UNSPSC Code:
50161901
Pricing and availability is not currently available.

agency

USP

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

assay

99.0-101.0%

autoignition temp.

698 °F

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol (name derived from Greek word glykys), a viscous liquid found in nature, can also be obtained from petrochemical raw materials. It is sweet in taste and commonly known as glycerine. The three hydroxyl groups are responsible for its solubility in water. It is used in the preparation of resins, emulsifiers, plasticizers, and stabilizers. It is also used as a solvent in many organic reactions.[1][2][3][4]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

390.2 °F - Pensky-Martens closed cup

flash_point_c

199 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycerol production and its applications as a raw material: A review
Tan H W, et al.
Renewable and Sustainable Energy Reviews, 27(24), 118-127 (2013)
Glycerol: Properties and Production
The Future of Glycerol (2008)
Glycerol as a sustainable solvent for green chemistry.
Gu Y and Jerome F.
Green Chemistry, 12(7), 1127-1138 (2010)
Mario Pagliaro et al.
Angewandte Chemie (International ed. in English), 46(24), 4434-4440 (2007-05-02)
Today, industrial plants that produce glycerol are closing down and others are opening that use glycerol as a raw material, owing to the large surplus of glycerol formed as a by-product during the production of biodiesel. Research efforts to find
Eric D B Goulet
International journal of sport nutrition and exercise metabolism, 19(5), 547-560 (2009-11-17)
Glycerol-induced hyperhydration (GIH) has been shown to improve fluid retention and endurance performance compared with water-induced hyperhydration. The goal of this article is to report on what is known and unknown about how glycerol-containing hyperhydration solutions (GCHSs) are processed at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service