Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001496

Sigma-Aldrich

Dimethyl sulfoxide

p.a., 99%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
506008
EC Number:
MDL number:
UNSPSC Code:
12191600
assay:
99%
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

grade

p.a.

assay

99%

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

dilution

(for analytical testing)

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongjie An et al.
Advances in colloid and interface science, 222, 9-17 (2014-08-17)
The observation by Atomic Force Microscopy of a range of nanophases on hydrophobic surfaces poses some challenging questions, not only related to the stability of these objects but also regarding their wetting properties. Spherical capped nanobubbles are observed to exhibit
A Oguro-Ando et al.
Molecular psychiatry, 20(9), 1069-1078 (2014-10-15)
Rare maternally inherited duplications at 15q11-13 are observed in ~1% of individuals with an autism spectrum disorder (ASD), making it among the most common causes of ASD. 15q11-13 comprises a complex region, and as this copy number variation encompasses many
Phillip McDonagh et al.
Veterinary microbiology, 174(3-4), 438-447 (2014-12-04)
Feline infectious peritonitis (FIP), a feline coronavirus (FCoV) induced disease, is almost invariably fatal with median life expectancy measured in days. Current treatment options are, at best, palliative. The objectives of this study were to evaluate a panel of nineteen
Claudia C Faria et al.
Cancer research, 75(1), 134-146 (2014-11-14)
Medulloblastoma is the most common malignant pediatric brain tumor, with metastases present at diagnosis conferring a poor prognosis. Mechanisms of dissemination are poorly understood and metastatic lesions are genetically divergent from the matched primary tumor. Effective and less toxic therapies
Rory W Blackler et al.
British journal of pharmacology, 172(4), 992-1004 (2014-10-10)
Hydrogen sulphide is an important mediator of gastrointestinal mucosal defence. The use of non-steroidal anti-inflammatory drugs (NSAIDs) is significantly limited by their toxicity in the gastrointestinal tract. Particularly concerning is the lack of effective preventative or curative treatments for NSAID-induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service