Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000376

Sigma-Aldrich

L-Valine

98.5-101.5%

Synonym(s):

(S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
1721136
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98.5-101.5%

form

crystalline

color

white

mp

295-300 °C (subl.) (lit.)

SMILES string

CC(C)[C@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1

InChI key

KZSNJWFQEVHDMF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hernan Carol et al.
Pediatric blood & cancer, 61(8), 1493-1496 (2014-03-26)
PF-03084014, a γ-secretase inhibitor, was tested against the PPTP in vitro cell line panel (1.0 nM to 10 μM) and against the in vivo xenograft panels (administered orally twice daily on Days 1-7 and 15-21). PF-03084014 demonstrated limited in vitro activity, with
B Leclercq
Poultry science, 77(1), 118-123 (1998-02-20)
Lysine exerts specific effects on body composition at dietary levels higher than that required for maximum growth rate, which also results in an improved feed conversion ratio (FCR). As a consequence of these differences, it becomes apparent that there is
Suang Suang Koid et al.
Hypertension (Dallas, Tex. : 1979), 63(4), 768-773 (2014-01-15)
Angiotensin-converting enzyme inhibitors and angiotensin AT1 receptor blockers reduce myocardial ischemia-reperfusion injury via bradykinin B2 receptor- and angiotensin AT2 receptor-mediated mechanisms. The renin inhibitor aliskiren increases cardiac tissue kallikrein and bradykinin levels. In the present study, we investigated the effect
Ijin Joo et al.
Radiology, 272(2), 417-426 (2014-04-05)
To evaluate the diagnostic value of intravoxel incoherent motion (IVIM) diffusion-weighted (DW) magnetic resonance (MR) imaging in the quantitative assessment of the therapeutic efficacy of a vascular disrupting agent (VDA) (CKD-516) in rabbit VX2 liver tumors. The institutional animal care
Xin Zhang et al.
Hypertension (Dallas, Tex. : 1979), 64(1), 87-93 (2014-04-23)
Renovascular hypertension alters cardiac structure and function. Autophagy is activated during left ventricular hypertrophy and linked to adverse cardiac function. The angiotensin II receptor blocker, valsartan, lowers blood pressure and is cardioprotective, but whether it modulates autophagy in the myocardium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service