Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I10009

Tetrahydrofuran

≥99.9%, suitable for HPLC, Vetec

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein/REAXYS Number:
102391
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
grade:
HPLC grade
assay:
≥99.9%
technique(s):
HPLC: suitable
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Tetrahydrofuran, suitable for HPLC, ≥99.9%

grade

HPLC grade

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

Vetec

assay

≥99.9%

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-6.2 °F - closed cup

flash_point_c

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Yan et al.
Advanced materials (Deerfield Beach, Fla.), 27(23), 3484-3491 (2015-05-06)
Polymer solar cells are fabricated with systematic variation of the phase purity. Photovoltaic tests demonstrate that devices with ca. 10% of mixed phases outperform pure-phase devices. Photophysical studies reveal the effects of mixed phase on charge generation and recombination. These
C N Ong et al.
British journal of industrial medicine, 48(9), 616-621 (1991-09-01)
Occupational exposure to tetrahydrofuran (THF) was studied by analysis of environmental air, blood, alveolar air, and urine from 58 workers in a video tape manufacturing plant. Head space gas chromatography (GC) with an FID detector was used for determination of
Ryan D Pensack et al.
Journal of the American Chemical Society, 137(21), 6790-6803 (2015-05-07)
We compare the singlet fission dynamics of five pentacene derivatives precipitated to form nanoparticles. Two nanoparticle types were distinguished by differences in their solid-state order and kinetics of triplet formation. Nanoparticles that comprise primarily weakly coupled chromophores lack the bulk
Luca Chiari et al.
The Journal of chemical physics, 138(7), 074301-074301 (2013-03-01)
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS)
P T Clayton et al.
The Journal of clinical investigation, 79(4), 1031-1038 (1987-04-01)
Urinary bile acids from a 3-mo-old boy with cholestatic jaundice were analyzed by ion exchange chromatography and gas chromatography-mass spectrometry (GC-MS). This suggested the presence of labile sulfated cholenoic acids with an allylic hydroxyl group, a conclusion supported by analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service