Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I10002

Hexane

suitable for HPLC, ≥99%

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein/REAXYS Number:
1730733
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

Vetec

assay

≥99%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-7.6 °F - closed cup

flash_point_c

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matti Marklund et al.
Circulation, 132(7), 586-594 (2015-06-19)
High intake of polyunsaturated fatty acids (PUFAs) may reduce the risk of cardiovascular disease (CVD) and mortality. Large, prospective studies including both sexes and circulating PUFAs as dietary biomarkers are needed. We investigated sex-specific associations of the major dietary PUFAs
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of
Alexandre S Cristino et al.
Nature communications, 5, 5529-5529 (2014-11-21)
Increasing evidence suggests small non-coding RNAs (ncRNAs) such as microRNAs (miRNAs) control levels of mRNA expression during experience-related remodelling of the brain. Here we use an associative olfactory learning paradigm in the honeybee Apis mellifera to examine gene expression changes
Jingjing Jin et al.
Journal of experimental botany, 66(13), 3959-3975 (2015-05-10)
The pleasant fragrance of ylang ylang varieties (Cananga odorata) is mainly due to volatile organic compounds (VOCs) produced by the flowers. Floral scents are a key factor in plant-insect interactions and are vital for successful pollination. C. odorata var. fruticosa
Elisabeth Christiansen et al.
The British journal of nutrition, 113(11), 1677-1688 (2015-04-29)
Various foods are associated with effects against metabolic diseases such as insulin resistance and type 2 diabetes; however, their mechanisms of action are mostly unclear. Fatty acids may contribute by acting as precursors of signalling molecules or by direct activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service