Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1645006

USP

Testolactone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Homo-17a-oxaandrosta-1,4-diene-3,17-dione

Sign Into View Organizational & Contract Pricing

Select a Size

125 MG
$221.00

$221.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
125 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$221.00


0 left in stock

Request a Bulk Order

grade

pharmaceutical primary standard

API family

testolactone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4(C(=CC(=O)C=C4)CC3)C)CC2)CCC1=O)C

InChI

1S/C19H24O3/c1-18-9-7-13(20)11-12(18)3-4-14-15(18)8-10-19(2)16(14)5-6-17(21)22-19/h7,9,11,14-16H,3-6,8,10H2,1-2H3/t14-,15+,16+,18+,19+/m1/s1

InChI key

BPEWUONYVDABNZ-DZBHQSCQSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Testolactone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service