Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1374995

USP

Mebendazole Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mebendazole-amine, 2-Amino-5-benzoyl-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3O
CAS Number:
Molecular Weight:
237.26
Beilstein/REAXYS Number:
3550536
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3

InChI

1S/C14H11N3O/c15-14-16-11-7-6-10(8-12(11)17-14)13(18)9-4-2-1-3-5-9/h1-8H,(H3,15,16,17)

InChI key

GPMHHSJZGVOEFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service