1265005
USP
Ethoxzolamide
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
6-Ethoxy-2-benzothiazolesulfonamide, 6-Ethoxyzolamide, Ethoxzolamide
About This Item
Recommended Products
manufacturer/tradename
USP
mp
190-193 °C (lit.)
application(s)
pharmaceutical (small molecule)
SMILES string
CCOc1ccc2nc(sc2c1)S(N)(=O)=O
InChI
1S/C9H10N2O3S2/c1-2-14-6-3-4-7-8(5-6)15-9(11-7)16(10,12)13/h3-5H,2H2,1H3,(H2,10,12,13)
InChI key
OUZWUKMCLIBBOG-UHFFFAOYSA-N
Gene Information
human ... CA1(759) , CA12(771) , CA13(377677) , CA14(23632) , CA2(760) , CA3(761) , CA4(762) , CA5A(763) , CA5B(11238) , CA6(765) , CA7(766) , CA9(768)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service