Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1190904

USP

Dicyclohexyl

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Bicyclohexyl, Dicyclohexyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11C6H11
CAS Number:
Molecular Weight:
166.30
Beilstein/REAXYS Number:
1848266
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

autoignition temp.

473 °F

manufacturer/tradename

USP

refractive index

n20/D 1.478 (lit.)

bp

227 °C (lit.)

mp

3-4 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

C1CCC(CC1)C2CCCCC2

InChI

1S/C12H22/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

WVIIMZNLDWSIRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service