1162626
USP
Alpha-Damascone
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
α-Damascone, 1-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-2-buten-1-one, 2,6,6-Trimethyl-1-crotonyl-2-cyclohexene
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
damascone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CC1=CCCC(C)(C)C1C(/C=C/C)=O
InChI
1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5,7-8,12H,6,9H2,1-4H3/b7-5+
InChI key
CRIGTVCBMUKRSL-FNORWQNLSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
related product
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service