Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1154650

USP

Allyl cyclohexane valerate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Allyl cyclohexanevalerate, 2-Propenyl 5-cyclohexanepentanoate

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 0.5 ML
$347.00

$347.00


Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
3 X 0.5 ML
$347.00

About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$347.00


Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

grade

pharmaceutical primary standard

API family

allyl cyclohexane valerate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(CC=C)C(=O)CCCCC1CCCCC1

InChI

1S/C14H24O2/c1-2-12-16-14(15)11-7-6-10-13-8-4-3-5-9-13/h2,13H,1,3-12H2

InChI key

NDXGAVFJHHVIQN-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Allyl cyclohexane valerate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service