Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1084995

USP

Caffeic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$405.00

$405.00


Check Cart for Availability


Select a Size

Change View
15 MG
$405.00

About This Item

Linear Formula:
(HO)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1954563
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

$405.00


Check Cart for Availability

grade

pharmaceutical primary standard

manufacturer/tradename

USP

mp

211-213 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+

InChI key

QAIPRVGONGVQAS-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

A natural dietary phenolic compound found in plants that is an anti-oxidant. Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.
Anti-oxidant phenolic compound found in plants; Inhibits the synthesis of leukotrienes.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service