1036008
USP
Anileridine hydrochloride
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Anileridine dihydrochloride, 1-[2-(4-Aminophenyl)ethyl]-4-phenyl-4-piperidinecarboxylic acid ethyl ester dihydrochloride
About This Item
grade
pharmaceutical primary standard
manufacturer/tradename
USP
drug control
USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)
application(s)
pharmaceutical (small molecule)
format
neat
InChI
1S/C22H28N2O2.2ClH/c1-2-26-21(25)22(19-6-4-3-5-7-19)13-16-24(17-14-22)15-12-18-8-10-20(23)11-9-18;;/h3-11H,2,12-17,23H2,1H3;2*1H
InChI key
ZYTHLJLPPSSDIP-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service