Key Documents
Z227269
Supelco® PEEK Fitting
Two-piece with ferrule, pkg of 5 ea
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
packaging
pkg of 5 ea
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Two-Piece Fingertight Fittings: Nut has wings to make fingertightening easier. Holds to 6000 psi (420 kg/cm2). Ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)