Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z226998

Spherisorb® C8 Guard Cartridge

5 μm particle size, L × I.D. 1 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

agency

suitable for USP L7

product line

Spherisorb®

feature

endcapped

packaging

pkg of 4 ea

extent of labeling

5.75% Carbon loading

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix active group

C8 (octyl) phase

particle size

5 μm

compatibility

for use with 3.0 to 4.6 mm column

separation technique

reversed phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service