Key Documents
Z226874
Supelco® PEEK Fitting
Hex-head nut, short, wrenchtight
About This Item
UNSPSC Code:
41115700
packaging
pkg of 5 ea
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Two-Piece Wrenchtight Fittings: Have a special double-tapered ferrule which is leak-free to 10,000 psi (700 kg/cm2). Hexagonal head. Double-tapered ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)