Key Documents
Z226564
Endfitting for HPLC hardware
Synonym(s):
HPLC column frits and end fittings
About This Item
UNSPSC Code:
41115700
material
316 stainless steel
fitting
1/4 in. female Valco compatible column side for 1/4 in. O.D. column body
1/16 in. female Valco compatible tubing side for 1/16 in. O.D. tubing
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)