Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002229

Atorvastatin for system suitability

Atorvastatin for system suitability
1 of 1 reviewers received a sample product or took part in a promotion

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Atorvastatin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H35FN2O5
CAS Number:
Molecular Weight:
558.64
UNSPSC Code:
41116107

API family

atorvastatin

manufacturer/tradename

EDQM

storage temp.

2-8°C

InChI

1S/C33H35FN2O5/c1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40/h3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40)/t26-,27-/m1/s1

InChI key

XUKUURHRXDUEBC-KAYWLYCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. North Carolina
    • Review 1
    • Votes 0
    2 out of 5 stars.

    Need better Results with EP Standards

    Needed this EP Standard, ordered in August and the batch1 expiry was Oct 2022 and the European Pharmacopoeia was already 6 months into selling batch 2. When trying to get Batch 2 I was told that the product has now been discontinued

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! We are sorry to hear that your experience did not meet expectations. We would like to learn more about your needs to see if there is a replacement product we can provide. When you have a moment, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Report Product Issue ticket. We look forward to hearing from you soon!

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service