Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TPSC18

Supelco

Supel-Tips C18 Pipette Tips

C18 bonded on spherical silica, endcapped, volume 10 μL, pkg of 96 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
volume:
10 μL
packaging:
pkg of 96 ea

packaging

pkg of 96 ea

volume

10 μL

particle size

50-60 μm

pore size

200 Å particle size

capacity

17 μg binding capacity (Insulin, Chain B, Oxidized)
18 μg binding capacity (Beta-Amayloid)
7.6 μg binding capacity (Bradykinin, Fragment 1-7)

application(s)

food and beverages

General description

Supel-Tips C18 pipette tip provides superior sample recovery, increased bed integrity, and enhanced binding capacity for purifying and concentrating mass spectrometry samples. Supel-Tips′ unique, proprietary adhesive chemistry greatly increases available silica particle surface area, leading to exceptional binding capacity and unsurpassed analyte recovery. The unique solid phase support binds peptides, proteins, and other analytes retained by reversed phase techniques while withstanding the rigors of repeated draw and dispense cycles without structural degradation. For optimal purification and concentration of important samples, trust Supel-Tips TPSC18.

Application

Recovery of peptides, proteins and other analytes retained by reversed phase techniques.

Features and Benefits

Discover the Advantages for Yourself!
  • Superior recovery of peptides and proteins
  • Exceptional binding capacity and enhanced affinity
  • Greater sorbent bed stability for cleaner samples

Other Notes

Browse all pipette tips

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service