Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TAN2

Supelco

TAN in Mineral Oil

analytical standard, set of 2 × 500 mL

Synonym(s):

TOTAL ACID NUMBER STANDARDS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

analytical standard

agency

ASTM® D664
according to ISO 17025
according to ISO 17034

packaging

set of 2 × 500 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
petroleum

format

multi-component solution

storage temp.

room temp

General description

Total Acid Number in Mineral Oil.

Standard No. 1 . . . . . Acid Number 2
Standard No. 2 . . . . . Acid Number 5

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service