Key Documents
SU861043
Supelco
Super Clean (Base-Plate Design) Kit
GC-FID fuel gas kit (includes SU861026, 2 X SU861025, and SU861013)
About This Item
UNSPSC Code:
41115717
NACRES:
SB.54
General description
Super Clean base-plate purifiers are a unique point-of use glass/metal, diffusion proof purification system to purifiy carrier, fuel, and other gases for the GC or GC-MS system. These purifiers remove hydrocarbons, oxygen (color indicated), and moisture (color indicated) to better than 6.0 gas (99.9999%) quality at 2 L/min., independent of the original gas quality.
Legal Information
Super Clean is a trademark of Scientific Glass Technology B.V.
Choose from one of the most recent versions:
Certificates of Analysis (COA)