Key Documents
SU860165
Supelco
Push-On cap
diam. 11 mm, blue polyethylene
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
material
blue polyethylene
feature
push-on cap
thinned penetration point, easy to pierce
packaging
pkg of 1 × 1000 ea (SU860165)
availability
not available in USA
diam.
11 mm
compatibility
for use with 11 mm crimp neck vials
Choose from one of the most recent versions:
Certificates of Analysis (COA)