SU860087
Vial crimp seals with PTFE/silicone/PTFE septa
seal diam. × thickness 8 mm × 1.0 mm, silver aluminum seal, red PTFE/silicone/red PTFE septum, pkg of 100 ea
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NB.11
Recommended Products
material
red PTFE/silicone/red PTFE septum
silver aluminum seal
hardness
45 (Durometer: shore A)
packaging
pkg of 100 ea
O.D.
8 mm
seal diam. × thickness
8 mm × 1.0 mm
Choose from one of the most recent versions:
Certificates of Analysis (COA)