Key Documents
SU860021
Supelco
Assembled screw cap with hole and PTFE/butyl septa
black polypropylene (center hole 5.5 mm), thread for 8-425, red PTFE/butyl rubber septum, pkg of 100 ea
About This Item
UNSPSC Code:
12352200
NACRES:
NB.11
Pricing and availability is not currently available.
material
black polypropylene (center hole 5.5 mm)
red PTFE/butyl rubber septum
hardness
55 (Durometer: shore A)
packaging
pkg of 100 ea
availability
not available in USA
thickness
1.3 mm
fitting
thread for 8-425
Choose from one of the most recent versions:
Certificates of Analysis (COA)