Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS90

Diazinon

analytical standard

Synonym(s):

Dimpylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N2O3PS
CAS Number:
Molecular Weight:
304.35
Beilstein/REAXYS Number:
273790
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

manufacturer/tradename

Chem Service, Inc. PS-90

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)C

InChI

1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3

InChI key

FHIVAFMUCKRCQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

219.9 °F - closed cup

flash_point_c

104.4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Zhang et al.
Environmental toxicology and pharmacology, 34(3), 959-968 (2012-09-12)
Pesticide exposure has repeatedly been associated with cancers. However, molecular mechanisms are largely undetermined. In this study, we examined whether exposure to diazinon, a common organophosphate that has been associated with cancers, could induce DNA methylation alterations. We conducted genome-wide
Karen Huen et al.
Environmental research, 117, 8-16 (2012-06-12)
Organophosphate pesticides are widely used and recent studies suggest associations of in utero exposures with adverse birth outcomes and neurodevelopment. Few studies have characterized organophosphate pesticides in human plasma or established how these levels correlate to urinary measurements. We measured
Dong Wang et al.
Reproductive toxicology (Elmsford, N.Y.), 34(4), 489-497 (2012-09-05)
The potential toxicity resulting from combinatorial effects of organophosphorus and pyrethroid insecticides are not completely known. We evaluated male reproductive toxicity in mice co-exposed to diazinon and cis-permethrin. Nine-week-old male Sv/129 mice were exposed to diazinon (10 μmol/kg/day) or cis-permethrin
M J van der Schans et al.
Chemico-biological interactions, 203(1), 96-102 (2012-11-06)
Organophosphate (OP) pesticides are neurotoxic compounds that are widely used in agriculture. Classical methods for monitoring OP exposure comprise the measurement of intact OP, its metabolites or cholinesterase activity. Newly developed methods focus on the analysis of the OP adduct
K P Shyma et al.
Experimental & applied acarology, 58(4), 471-481 (2012-07-05)
The multi host tick, Hyalomma anatolicum, is the commonest Hyalomma species in India and cattle serves as the main host of this species. A study to evaluate the acaricide resistance of H. anatolicum to deltamethrin, cypermethrin and diazinon was conducted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service