Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS902-250MG

N,N-Diethyl-m-toluamide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Beilstein/REAXYS Number:
2046711
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-902

SMILES string

CCN(CC)C(=O)c1cccc(C)c1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3

InChI key

MMOXZBCLCQITDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Robbins et al.
Journal of toxicology and environmental health, 18(4), 503-525 (1986-01-01)
A review of the biodistribution and toxicity of the insect repellent N,N-diethyl-m-toluamide (DEET) is presented. Workers using repellent containing this compound may be exposed to greater than 442 g in 6 mo. In human studies, variable penetration into the skin
Pinky Kain et al.
Nature, 502(7472), 507-512 (2013-10-04)
There are major impediments to finding improved DEET alternatives because the receptors causing olfactory repellency are unknown, and new chemicals require exorbitant costs to determine safety for human use. Here we identify DEET-sensitive neurons in a pit-like structure in the
Oxana A Belova et al.
Ticks and tick-borne diseases, 3(4), 240-246 (2012-08-23)
There is some evidence that tick-borne encephalitis virus (TBEV) prevalence in ticks, removed from humans, is higher than that in field-collected ticks from the same area. There are two possible explanations: (i) Infected ticks are more active and aggressive and
Nina M Stanczyk et al.
PloS one, 8(2), e54438-e54438 (2013-02-26)
DEET (N,N-Diethyl-m-toluamide) is one of the most widely used mosquito repellents. Although DEET has been shown to be extremely effective, recent studies have revealed that certain individual insects are unaffected by its presence. A genetic basis for this has been
J R Clem et al.
The Annals of pharmacotherapy, 27(3), 289-293 (1993-03-01)
To describe a case of N,N-diethyl-m-toluamide (DEET)-induced cardiovascular toxicity in an adult and reviews other cases that have been reported in the published literature. Human and animal data available on DEET pharmacokinetics are reviewed and factors that predispose an individual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service