Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS79-250MG

Toxaphene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-79

application(s)

cleaning products
cosmetics
food and beverages
personal care

InChI

1S/C10H8Cl8/c1-4-7(2-11,3-12)9(16)6(14)5(13)8(4,15)10(9,17)18/h5-6H,1-3H2

InChI key

OEJNXTAZZBRGDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Xia et al.
Analytical and bioanalytical chemistry, 395(2), 457-463 (2009-07-31)
Toxaphene is considered to be a problematic organochlorine pollutant because of its bioaccumulation potential and persistence in aquatic environments. In this study, whole lake trout and walleye composites were used to evaluate two analytical techniques for total toxaphene and selected
Seonock Woo et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 156(1), 37-41 (2012-04-14)
We evaluated toxaphene-induced acute toxicity in Hydra magnipapillata. The median lethal concentrations of the animals (LC(50)) were determined to be 34.5 mg/L, 25.0 mg/L and 12.0 mg/L after exposure to toxaphene for 24 h, 48 h and 72 h, respectively.
M H G Berntssen et al.
Aquatic toxicology (Amsterdam, Netherlands), 112-113, 54-61 (2012-03-01)
The relative feed-to-fish accumulation and possible biotransformation of the nona-chlorinated toxaphene congeners currently included in EU-legislation (CHB-50 and -62) and the octa-chlorinated congeners recommended by the European Food Safety Authority to be included in future surveillance of fish samples (CHB-40
Louise Champoux et al.
Environmental toxicology and chemistry, 29(2), 243-249 (2010-09-08)
The great blue heron (Ardea herodias) has been used as a bioindicator of the state of the St. Lawrence River (Québec, Canada) since 1996. At 5-year intervals, selected breeding colonies along the River and its estuary are visited to estimate
Seonock Woo et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 153(3), 355-361 (2011-01-12)
Differential gene expression profiles were established from the head and liver tissues of the marine medaka fish (Oryzias javanicus) after its exposure to toxaphene, a persistent organic pollutant and insecticide, using differential display polymerase chain reaction. Twenty-seven differentially expressed genes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service