Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS61-250MG

1,1-Dimethyl-3-phenylurea

analytical standard

Synonym(s):

1,1-Dimethyl-3-phenylurea, Fenuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O
CAS Number:
Molecular Weight:
164.20
Beilstein/REAXYS Number:
2208535
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-61

SMILES string

CN(C)C(=O)Nc1ccccc1

InChI

1S/C9H12N2O/c1-11(2)9(12)10-8-6-4-3-5-7-8/h3-7H,1-2H3,(H,10,12)

InChI key

XXOYNJXVWVNOOJ-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Richard et al.
Environmental science & technology, 38(7), 2052-2057 (2004-04-29)
The IHSS soil humic acid (HA) standard and two HAs from soils of very different origin (Chernozem and Ranker) were fractionated by tandem size-exclusion chromatography-polyacrylamide gel electrophoresis. From each HA, three fractions with different molecular sizes (MSs) and electrophoretic mobilities
Yen Hai Dao et al.
Water research, 45(11), 3309-3317 (2011-04-26)
The relative rates of degradation of three hydroxyl radical probe compounds (atrazine, fenuron and parachlorobenzoic acid (pCBA)) by Fe(III)/H(2)O(2) (pH = 2.85), Fe(III)NTA/H(2)O(2) (neutral pH), Fe(II)/O(2), Fe(II)NTA/O(2), Fe(II)/H(2)O(2) and Fe(II)NTA/H(2)O(2) (neutral pH) have been investigated using the competitive kinetic method.
[Embryotropic effect of fenuron].
L T Voloshina
Vrachebnoe delo, (2)(2), 103-105 (1985-02-01)
H H Attaway et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 17(6), 683-699 (1982-01-01)
Anaerobic degradation of diuron [3-(3,4-dichlorophenyl)-1,1-dimethylurea], monuron [3-(4-chlorophenyl)-1,1-dimethylurea], and fenuron [1,1-dimethyl-3-phenylurea] were studied. Herbicide containing media (reduced with cysteine-HCl and under 95% N2:5% CO2 gas phase) were inoculated with pond sediments. Sediment from a diuron-treated pond dehalongenated diuron to 3-(3-chlorophenyl)-1,1-dimethylurea (CPDU)
[Hygienic evaluation of the method of ozonization used for detoxication of aqueous solutions of urea derivatives].
V I Tsipriian et al.
Gigiena i sanitariia, (2)(2), 83-85 (1985-02-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service