Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS56

Dinoseb

analytical standard

Synonym(s):

2-sec-Butyl-4,6-dinitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O5
CAS Number:
Molecular Weight:
240.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-56

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCC(C)c1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C10H12N2O5/c1-3-6(2)8-4-7(11(14)15)5-9(10(8)13)12(16)17/h4-6,13H,3H2,1-2H3

InChI key

OWZPCEFYPSAJFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

306.9 °F - closed cup

flash_point_c

152.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Satapanajaru et al.
Journal of hazardous materials, 168(2-3), 930-937 (2009-04-07)
Dinoseb, a dinitroherbicide, was once commonly used in aerial crop dusting of agronomic crops in the western United States. Widespread use combined with improper disposal practices at rural air strips has contaminated numerous sites. Our objective was to determine if
Carine Staempfli et al.
Ecotoxicology and environmental safety, 68(2), 263-271 (2007-01-30)
This study evaluated the usefulness of energy reserves as an early warning system in Folsomia candida when exposed to the pesticide dinoseb. After different exposure times, survival, reproduction, growth (weight and length), lipid and protein content of the organisms were
Nathalie Chèvre et al.
Ecotoxicology and environmental safety, 62(1), 17-25 (2005-06-28)
Models describing dose-response relationships are becoming increasingly popular in ecotoxicology. They allow simple and thorough evaluations of toxicity test results, including inter- and extrapolations to concentrations or exposure times other than those tested. Simple parametric regression models are of particular
Mallipattu Sreedhar et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(4), 511-516 (2003-05-03)
A sensitive adsorptive stripping voltammetric method for the determination of dinitrophenolic herbicides, dinoseb (DSB) and dinoterb (DTB) at a bare carbon paste electrode (CPE) and a clay modified carbon paste electrode (CMCPE) was developed. A systematic study of various experimental
Mark R Viant et al.
Aquatic toxicology (Amsterdam, Netherlands), 77(4), 359-371 (2006-02-21)
Pesticide pulses in the Sacramento River, California, originate from storm-water discharges and non-point source aquatic pollution that can last from a few days to weeks. The Sacramento River and its tributaries have historically supported the majority of California's Chinook salmon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service