Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS380-250MG

Atrazine

analytical standard

Synonym(s):

2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
Beilstein/REAXYS Number:
612020
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-380

SMILES string

CCNc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)

InChI key

MXWJVTOOROXGIU-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Graymore et al.
Environment international, 26(7-8), 483-495 (2001-08-04)
A portion of all herbicides applied to forests, croplands, road sides, and gardens are inevitably lost to water bodies either directly through runoff or indirectly by leaching through groundwater into ephemeral streams and lakes. Once in the aquatic environment, herbicides
V Migeot et al.
Environmental research, 122, 58-64 (2013-01-24)
Groundwater, surface water and drinking water are contaminated by nitrates and atrazine, an herbicide. They are present as a mixture in drinking water and with their endocrine-disrupting activity, they may alter fetal growth. To study an association between drinking-water atrazine
Tyrone B Hayes et al.
The Journal of steroid biochemistry and molecular biology, 127(1-2), 64-73 (2011-03-23)
Atrazine is the most commonly detected pesticide contaminant of ground water, surface water, and precipitation. Atrazine is also an endocrine disruptor that, among other effects, alters male reproductive tissues when animals are exposed during development. Here, we apply the nine
Derek W Gammon et al.
Pest management science, 61(4), 331-355 (2005-01-19)
A risk assessment of the triazine herbicide atrazine has been conducted by first analyzing the toxicity database and subsequently estimating exposure. Margins of safety (MOS) were then calculated. Toxicity was assessed in animal studies and exposure was estimated from occupational
T Komang Ralebits et al.
Biodegradation, 13(1), 11-19 (2002-09-12)
The potential toxicity of the s-triazine herbicide atrazine motivates continuous bioremediation-directed research. Several indigenous soil atrazine-catabolizing microbial associations and monocultures have been enriched/isolated from compromised sites. Of these, Pseudomonas sp. strain ADP has become a reference strain and has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service