Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS342

Silvex® methyl ester

ampule of 250 mg

Synonym(s):

Methyl 2-(2,4,5-trichlorophenoxy)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9Cl3O3
CAS Number:
Molecular Weight:
283.54
Beilstein/REAXYS Number:
2135850
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-342

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

COC(=O)C(C)Oc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C10H9Cl3O3/c1-5(10(14)15-2)16-9-4-7(12)6(11)3-8(9)13/h3-5H,1-2H3

InChI key

YTAXYXOJOYIQQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Silvex is a registered trademark of Silberline Manufacturing Co., Inc.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service