Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS293-250MG

Dichloran

analytical standard

Synonym(s):

2,6-Dichloro-4-nitroaniline, Dichloran

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
Beilstein/REAXYS Number:
1459581
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-293

mp

190-192 °C (lit.)

application(s)

agriculture
environmental

SMILES string

Nc1c(Cl)cc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2

InChI key

BIXZHMJUSMUDOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florina Uzefovsky et al.
Emotion (Washington, D.C.), 14(4), 712-721 (2014-05-29)
Increasing evidence points to a role of dopaminergic pathways in modulating social behavior. Specifically, a polymorphic region in the third exon of the Dopamine D4 receptor (DRD4) has been associated with a host of social behaviors, often in an environment-sensitive
S Andrews et al.
Applied and environmental microbiology, 51(6), 1235-1238 (1986-06-01)
A selective medium containing 2 micrograms of dichloran per ml, 200 micrograms of chloramphenicol per ml, and 1.5% bacteriological peptone was developed for the isolation of Fusarium species and dematiaceous hyphomycetes from cereals. The medium, designated DCPA, was shown to
S M Schrap et al.
Chemosphere, 40(12), 1389-1397 (2000-05-02)
To be able to predict the degradation (rate) of organic chemicals (e.g. pesticides) in the field, knowledge of the environmental conditions that are of influence on the degradation process are of importance. In the present study an experimental system is
A more selective medium for Culicinomyces clavisporus.
C Panter et al.
Journal of invertebrate pathology, 82(3), 198-200 (2003-04-05)
L Gómez-Gómez et al.
Plant molecular biology, 30(4), 821-832 (1996-02-01)
Two cDNA clones coding for S-adenosyl-L-methionine synthase (SAMs, EC 2.5.1.6) have been isolated from a cDNA library of gibberellic acid-treated unpollinated pea ovaries. Both cDNAs were sequenced showing a high degree of identity but coding for different SAMs polypeptides. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service