Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS221-250MG

Metam sodium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS221-250MG

InChI

1S/C2H5NS2.Na/c1-3-2(4)5;/h1H3,(H2,3,4,5);/q;+1/p-1

InChI key

AFCCDDWKHLHPDF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren Gay et al.
Avian diseases, 54(1 Suppl), 763-767 (2010-06-05)
The chemical compound metam-sodium was tested at three concentrations for the ability to inactivate the infectivity of low-pathogenic avian influenza virus (LPAIV) and infectious bursal disease virus (IBDV), with virus-contaminated chicken litter used as the substrate. LPAIV was inactivated within
R H Fetterer et al.
The Journal of parasitology, 96(3), 632-637 (2010-06-19)
Metam sodium (MS, sodium N-methyldithiocarbamate) is a widely used soil pesticide. Fumigation or chemical sterilization of poultry litter containing infectious oocysts could be an effective strategy to block the transmission of avian coccidia. In the current study, the effect of
Presidential address. Sanitizing sewage sludge: the intersection of parasitology, civil engineering, and public health.
George D Cain
The Journal of parasitology, 96(6), 1037-1040 (2010-12-17)
M Díaz-Pérez et al.
Microbial ecology, 57(2), 379-383 (2008-11-11)
The monoculture of melon in Guatemala has caused the massive appearance of plants with an analogous syndrome for the well-known disease commonly called melon collapse, or vine decline, causing significant losses in crops. Methyl bromide is commonly used to sterilize
Seyed N Azizi et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 45(8), 766-773 (2010-10-23)
This study employs the Taguchi optimization methodology to optimize the effective parameters for the pesticide (Vapam) sorption onto soil modified with natural zeolite (clinoptilolite). The experimental factors and their ranges chosen for determination of the effective parameters were: initial Vapam

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service