Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS2002

Fenpropathrin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO3
CAS Number:
Molecular Weight:
349.42
Beilstein/REAXYS Number:
2673776
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-2002

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2)C1(C)C

InChI

1S/C22H23NO3/c1-21(2)19(22(21,3)4)20(24)26-18(14-23)15-9-8-12-17(13-15)25-16-10-6-5-7-11-16/h5-13,18-19H,1-4H3

InChI key

XQUXKZZNEFRCAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanfan Hong et al.
Journal of environmental management, 91(11), 2295-2300 (2010-07-14)
A gram-negative fenpropathrin-degrading bacterial strain Sphingobium sp. JQL4-5 was isolated from the wastewater treatment sludge of an insecticide factory. Strain JQL4-5 showed the ability to degrade other pyrethroid insecticides, but it was not able to degrade methyl parathion. To enhance
S Sivasupramaniam et al.
Journal of economic entomology, 93(3), 949-954 (2000-07-21)
A large, genetically diverse pool of Bemisia argentifolii Bellows & Perring was collected in 1994 from different crops, and a mixed colony was established in the laboratory. Subsets of this colony were reared on cotton plants held in large Plexiglas
A Maklakov et al.
Journal of economic entomology, 94(5), 1059-1066 (2001-10-30)
The fruit fly Dacus ciliatus Loew is a pest of the fruits of many cucurbit species. We studied the effect of organaophosphate and pyrethroid compounds on the adult flies by using surface contact and oral administration. In contrast to other
Zhaoyany Li et al.
Chemosphere, 76(4), 509-516 (2009-04-10)
Fenpropathrin and fenvalerate are two widely used pyrethroid insecticides. Due to one and two asymmetrical centers, fenpropathrin and fenvalerate consist of two and four stereoisomers, respectively. In the present study, the degradation of fenpropathrin and fenvalerate in two soils, an
J-Z Niu et al.
Insect molecular biology, 21(4), 422-436 (2012-06-09)
The citrus red mite, Panonychus citri, is known for its ability rapidly to evolve resistance to insecticides/acaricides and to adapt to hosts that produce toxins. In this study, we constructed an unprecedented four gigabase pair transcriptome of P. citri, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service