Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS1033-250MG

Oxyfluorfen

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Beilstein/REAXYS Number:
2065259
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-1033

SMILES string

CCOc1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C15H11ClF3NO4/c1-2-23-14-8-10(4-5-12(14)20(21)22)24-13-6-3-9(7-11(13)16)15(17,18)19/h3-8H,2H2,1H3

InChI key

OQMBBFQZGJFLBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamdy M A Hassanein
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 37(4), 521-527 (2002-06-06)
The alterations of the AChE activity in the brains of two fresh water fishes; Oreochromis niloticus and Gambusia affinis were measured after exposure to acute, sub-acute and chronic concentrations from the widely used herbicide; oxyfluorfen. Bioassays were conducted under controlled
Chloé Bonnineau et al.
Ecotoxicology (London, England), 21(4), 1208-1224 (2012-03-13)
In multiple stress situations, the co-occurrence of environmental and chemical factors can influence organisms' ability to cope with toxicity. In this context, the influence of light adaptation on the response of freshwater biofilms to sudden light changes or to herbicides
Laura Scrano et al.
Journal of environmental quality, 33(2), 605-611 (2004-04-13)
The photochemical behavior of oxyfluorfen [2-chloro-1-(3-etoxy-4-nitrophenoxy)-4-(trifluoromethyl) benzene] on two Greek soils was investigated. Soils were sampled from Nea Malgara and Preveza regions, characterized by a different organic matter content. Soils were spiked with the diphenyl-ether herbicide and irradiation experiments were
Iris Sandorf et al.
Planta, 216(1), 173-179 (2002-11-14)
Coronatine-inducible tyrosine aminotransferase (TAT), which catalyses the transamination from tyrosine to p-hydroxyphenylpyruvate, is the first enzyme of a pathway leading via homogentisic acid to plastoquinone and tocopherols, the latter of which are known to be radical scavengers in plants. TAT
Sheeba et al.
Ecotoxicology and environmental safety, 74(7), 1981-1993 (2011-07-30)
In the present study, degree of tolerance and tolerance strategies of two paddy field cyanobacteria viz. Nostoc muscorum and Phormidium foveolarum against oxyfluorfen (10 and 20 μg ml(-1)) and UV-B (7.2 kJ m(-2)d(-1)) stress were investigated. Oxyfluorfen and UV-B decreased

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service