Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS1033-100MG

Oxyfluorfen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Beilstein/REAXYS Number:
2065259
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1033

application(s)

agriculture
environmental

SMILES string

CCOc1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C15H11ClF3NO4/c1-2-23-14-8-10(4-5-12(14)20(21)22)24-13-6-3-9(7-11(13)16)15(17,18)19/h3-8H,2H2,1H3

InChI key

OQMBBFQZGJFLBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sushil K Kothari et al.
Pest management science, 58(12), 1254-1258 (2002-12-13)
Abstract: Field investigations were carried out during 1999 and 2000 to identify effective chemical/ cultural methods of weed control in rose-scented geranium (Pelargonium spp). The treatments comprised pre-emergence applications of oxyfluorfen (0.15, 0.20 and 0.25 kg AI ha(-1)) and pendimethalin
H M Hassanein et al.
Archives of environmental contamination and toxicology, 37(1), 78-84 (1999-05-26)
This paper deals with the expression of the biomarker hsp70 in the liver and kidney of the freshwater fish Oreochromis niloticus following exposure to the herbicide oxyfluorfen (Goal). Fishes were exposed to three concentrations, the 96-h LC50 (3 mg/L), the
L Geoffroy et al.
Archives of environmental contamination and toxicology, 45(4), 445-452 (2004-01-08)
The effect of oxyfluorfen was investigated when alga Scenedesmus obliquus has been exposed to different concentrations (7.5, 15, and 22.5 microg x L(-1)) at 12, 24, and 48 hours of exposure. Toxicity test was done by using 13 biomarkers concerning
Eunjoo Jeong et al.
Bioorganic chemistry, 31(5), 389-397 (2003-08-28)
In an effort to asses the effect of Val311Met point mutation of Bacillus subtilis protoporphyrinogen oxidase on the resistance to diphenyl ether herbicides, a Val311Met point mutant of B. subtilis protoporphyrinogen oxidase was prepared, heterologously expressed in Escherichia coli, and
Shobha Sondhia
Environmental monitoring and assessment, 162(1-4), 163-168 (2009-02-25)
A field study was conducted to determine persistence and bioaccumulation of oxyflorfen residues in onion crop at two growth stages. Oxyfluorfen (23.5% EC) was sprayed at 250 and 500 g ai/ha on the crop (variety, N53). Mature onion and soil

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service