Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLPE3-5EP-25GM

Standards for PE Analysis

Synonym(s):

Polyethylene Standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

packaging

set of 3 × standards

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

RoHS/WEEE complaint polymer standards in powder form for analysis of materials in polyethylene (PE). One Quality Control Sample comes with the 9 standard sets only.

  • Standard No. 1 (Wt %)
  • Br 0.0000
  • Cd 0.0000
  • Cr 0.0000
  • Hg 0.0000
  • Pb 0.0000

  • Standard No. 2 (Wt %)
  • Br 0.0250
  • Cd 0.0050
  • Cr 0.0500
  • Hg 0.0500
  • Pb 0.0500

  • Standard No. 3 (Wt %)
  • Br 0.0500
  • Cd 0.0100
  • Cr 0.1000
  • Hg 0.1000
  • Pb 0.1000

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service