Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL85758

11-Methylsulfinylundecylglucosinolate potassium salt

phyproof® Reference Substance

Synonym(s):

Potassium 11-methylsulfinylundecylglucosinolate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$947.00

$947.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$947.00

About This Item

Empirical Formula (Hill Notation):
C19H36KNO10S3
CAS Number:
Molecular Weight:
573.78
UNSPSC Code:
41116107
NACRES:
NA.24

$947.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

OC[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H](S/C(CCCCCCCCCCCS(C)=O)=N/OS(=O)(O[K])=O)O1

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service