Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL85723

Chamaemeloside

phyproof® Reference Substance

Synonym(s):

Apigenin 7-[6′′-(3-hydroxy-3-methylglutaryl)glucoside]

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$508.00

$508.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$508.00

About This Item

Empirical Formula (Hill Notation):
C27H28O14
CAS Number:
Molecular Weight:
576.50
UNSPSC Code:
41116107
NACRES:
NA.24

$508.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C(CC(O)(C)CC(O)=O)OC[C@@H](O1)[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1OC2=CC(O)=C(C(C=C(C3=CC=C(O)C=C3)O4)=O)C4=C2

InChI

1S/C27H28O14/c1-27(37,9-20(31)32)10-21(33)38-11-19-23(34)24(35)25(36)26(41-19)39-14-6-15(29)22-16(30)8-17(40-18(22)7-14)12-2-4-13(28)5-3-12/h2-8,19,23-26,28-29,34-37H,9-11H2,1H3,(H,31,32)/t19-,23-,24+,25-,26-,27?/m1/s1

InChI key

WWGLAVUKYJELNJ-KJTOLNCNSA-N

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service