Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL84091

Heliosupine

phyproof® Reference Substance

Synonym(s):

Cyanoglossophine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO7
CAS Number:
Molecular Weight:
397.46
UNSPSC Code:
41116107

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

InChI

1S/C20H31NO7/c1-6-12(2)17(23)28-15-8-10-21-9-7-14(16(15)21)11-27-18(24)20(26,13(3)22)19(4,5)25/h6-7,13,15-16,22,25-26H,8-11H2,1-5H3/b12-6-/t13-,15-,16+,20+/m0/s1

InChI key

HRSGCYGUWHGOPY-UKLMUADPSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Analysis Note

For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakob A Shimshoni et al.
Molecules (Basel, Switzerland), 26(3) (2021-02-03)
Pyrrolizidine alkaloids (PAs) are genotoxic carcinogenic phytotoxins mostly prevalent in the Boraginaceae, Asteraceae and Fabaceae families. Heliotropium species (Boraginaceae) are PA-producing weeds, widely distributed in the Mediterranean region, that have been implicated with lethal intoxications in livestock and humans. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service