Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL84074

Sceleratine N-oxide

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$535.00

$535.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$535.00

About This Item

Empirical Formula (Hill Notation):
C18H27NO8
CAS Number:
Molecular Weight:
385.41
UNSPSC Code:
41116107
NACRES:
NA.24

$535.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Check out our entire range of pyrrolizidine alkaloid standards

Analysis Note

For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service