Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL82633

Pseudolaric acid A-glucopyranoside

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$536.00

$536.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$536.00

About This Item

Empirical Formula (Hill Notation):
C28H38O11
CAS Number:
Molecular Weight:
550.59
UNSPSC Code:
41116107
NACRES:
NA.24

$536.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC(=O)\C(=C\C=C\[C@]2(OC(=O)[C@]43[C@@]([C@H]2CC4)(CCC(=CC3)C)OC(=O)C)C)\C

InChI

1S/C28H38O11/c1-15-7-11-27-12-9-19(28(27,13-8-15)38-17(3)30)26(4,39-25(27)35)10-5-6-16(2)23(34)37-24-22(33)21(32)20(31)18(14-29)36-24/h5-7,10,18-22,24,29,31-33H,8-9,11-14H2,1-4H3/b10-5+,16-6+/t18-,19+,20-,21+,22-,24+,26-,27-,28+/m1/s1

InChI key

IVYWRYGMQNKDQB-VHJBJYHKSA-N

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service