Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL82487

Berbamine dihydrochloride

phyproof® Reference Substance

Synonym(s):

berbenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H40N2O6 ·2HCl
CAS Number:
Molecular Weight:
681.65
MDL number:
UNSPSC Code:
41116107

grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥90.0% (HPLC)

optical activity

[α]20/D +116°, c = 1% in ethanol

manufacturer/tradename

PhytoLab

mp

250-253 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

Cl.Cl.COc1cc2CCN(C)[C@H]3Cc4ccc(Oc5cc(C[C@H]6N(C)CCc7cc(OC)c(OC)c(Oc1cc23)c67)ccc5O)cc4

InChI

1S/C37H40N2O6.2ClH/c1-38-14-12-24-19-32(41-3)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-31-18-23(8-11-30(31)40)17-29-35-25(13-15-39(29)2)20-34(42-4)36(43-5)37(35)45-33;;/h6-11,18-21,28-29,40H,12-17H2,1-5H3;2*1H/t28-,29+;;/m0../s1

InChI key

USRXDYNDPPUBSG-KKXMJGKMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

Reactant for:
  • Preparation of and anti-leukemia active berbamine derivatives

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankita Sharma et al.
Food & function, 12(2), 892-909 (2021-01-08)
Non-alcoholic fatty liver disease (NAFLD), a chronic metabolic disorder is concomitant with oxidative stress and inflammation. This study aimed to assess the effects of berbamine (BBM), a natural bisbenzylisoquinoline alkaloid with manifold biological activities and pharmacological effects on lipid, cholesterol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service