Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OC9-1KT

Supelco

Fatty Acids, Odd Carbon Straight Chains Kit

(individually packaged in quantities indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 1 kit

concentration

(individually packaged in quantities indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Heneicosanoic acid 1 g
    Heptadecanoic acid 1 g
    Heptanoic acid 1 g
    Nonadecanoic acid 1 g
    Nonanoic acid 1 g
    Pentadecanoic acid 1 g
    Tricosanoic acid 1 g
    Tridecanoic acid 1 g
    Undecanoic acid 1 g
    See All (9)

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

235.4 °F

flash_point_c

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service