Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NEFOR011

NEMo Sensor for Formaldehyde

Synonym(s):

Profil`Air®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

availability

available only in EU

size

14 × 7 × 3

application(s)

environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

The combined analysis of these two parameters allows optimal management of the building′s energy performance, at the same time preserving the health of its occupants.

Features and Benefits

Formaldehyde sensors for NeMo logger. The sensors used the ETHERA′s patented technology.

Suitability

Fits the following instrument:
NEKIT420

Other Notes

Weight: 0.1
Temperature conditions: Cooler/Regrigerated

Legal Information

Profil'Air is a registered trademark of Ethera S.A.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service