Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N13142

Pyraclofos

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 10 mg

manufacturer/tradename

Chem Service, Inc. PS-2046

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

InChI

1S/C14H18ClN2O3PS/c1-3-9-22-21(18,19-4-2)20-14-10-16-17(11-14)13-7-5-12(15)6-8-13/h5-8,10-11H,3-4,9H2,1-2H3

InChI key

QHGVXILFMXYDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxin Xu et al.
Journal of agricultural and food chemistry, 60(17), 4173-4178 (2012-04-13)
This study investigated the enantioselective degradation behavior of pyraclofos in three soils (NC, HZ, and ZZ) under native and sterilized conditions. The absolute configuration of pyraclofos enantiomers has been determined by the combination of experimental and calculated electronic circular dichroism
S S Sahu
Indian journal of malariology, 33(1), 16-20 (1996-03-01)
A hut-scale trial of pyraclofos 50% EC applied as an indoor residual spray at 1 gm/sq m was carried out against malaria vectors. Anopheles fluviatilis and An. culicifacies in Kandhaguda village of Malkangiri district, Orissa. Bandhaguda village was kept as
Huayun Yang et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 43(3), 219-223 (2008-03-28)
The hydrolysis of the insecticide pyraclofos in buffered solutions at pH 5.0, 7.0 and 9.0, and its sorption on four soils of different physicochemical properties were investigated. The results showed that the degradation of pyraclofos in buffered solutions followed pseudo-first-order
Honghua Zhang et al.
Journal of agricultural and food chemistry, 60(28), 6953-6959 (2012-06-20)
Despite the fact that the biological processes of chiral pesticides are enantioselective, knowledge of the toxicities of pyraclofos due to enantiospecificity is scarce. In this study, the optical isomers of pyraclofos were separated and their toxicities to butyrylcholinesterase (BChE) and
R Velayudhan et al.
The Journal of communicable diseases, 22(2), 140-147 (1990-06-01)
Carbosulfan (OMS 3022), a carbamate and pyraclofos (OMS 3040), an organophosphate were evaluated for their biological activity against vector mosquitoes, both in laboratory and field conditions. Carbosulfan (25% EC), was found to be active against adults and immatures of Culex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service