Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N13136

Propyzamide

analytical standard

Synonym(s):

Pronamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Cl2NO
CAS Number:
Molecular Weight:
256.13
Beilstein/REAXYS Number:
882391
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 1 g

manufacturer/tradename

Chem Service, Inc. PS-349

storage temp.

−20°C

SMILES string

CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C

InChI

1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)

InChI key

PHNUZKMIPFFYSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyzamide is a systemic herbicide that belongs to the amides family. The herbicide acts by disturbing the mitotic sequence in dividing cells thus obstructing the growth of the target species. Due to its low toxicity, it is widely utilized as a pre-emergent and post-emergent herbicide for controlling weeds in several plants including alfalfa, lettuce, sunflower, etc.[1][2]

Application

Propyzamide may be used as an analytical standard for the determination of the analyte in air,[3] urban precipitation[4] and water samples[5] by chromatography based techniques.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Fenoll et al.
Talanta, 85(2), 975-982 (2011-07-06)
In this work, a rapid and sensitive analytical multiresidue method has been developed for the simultaneous determination of 48 pesticides and 19 metabolites in waters (tap, leaching and sewage), using liquid chromatography-tandem mass spectrometry (LC-MS/MS) with triple quadrupole in selected
Simultaneous determination of trace pesticides in urban precipitation
Haraguchi K, et al.
Atmospheric Environment, 29(2), 247-253 (1995)
Baiping Zhao et al.
Bioresource technology, 179, 144-149 (2014-12-30)
Propyzamide is a widely used benzamide herbicide for controlling weeds in lettuce, soybeans, cotton and other crops. An efficient propyzamide-degrading strain W1 was firstly isolated from activated sludge and identified as Comamonas testosteroni. A metabolite of propyzamide by strain W1
Simple GC and GC/MS determination of Herbicides in air using quartz fiber filter and activated carbon fiber filter for sampling
Kawata K
Journal of Environmental Chemistry, 2(2), 181-186 (1992)
Yoshiteru Mizukoshi et al.
Ultrasonics sonochemistry, 12(3), 191-195 (2004-10-20)
We prepared Au/gamma-Fe2O3 composite nanoparticles by sonochemically reducing Au(III) ions employing no stabilizer in the aqueous solution to form stable Au nanoparticles and allowing them to attach onto the surface of gamma-Fe2O3 particles with an average size of 21 nm.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service