Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N12831

Pentachlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Cl5OH
CAS Number:
Molecular Weight:
266.34
Beilstein/REAXYS Number:
1285380
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

9.2 (vs air)

vapor pressure

40 mmHg ( 211.2 °C)

packaging

ampule of 1 g

manufacturer/tradename

Chem Service, Inc. PS-7

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

310 °C (lit.)

mp

165-180 °C (lit.)

density

1.978 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

IZUPBVBPLAPZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Di Leo et al.
Journal of hazardous materials, 244-245, 303-310 (2012-12-29)
The existence of a lot of worldwide pentachlorophenol-contaminated sites has induced scientists to concentrate their effort in finding ways to degrade it. Therefore, an effective tool to decompose it from soil mixtures is needed. In this work the efficiency of
J P Seiler
Mutation research, 257(1), 27-47 (1991-01-01)
Pentachlorophenol (PCP) is a substance whose widespread use, mainly in wood protection and pulp and paper mills, has led to a substantial environmental contamination. This in turn accounts for a significant exposure of the general human population, with rather high
Weiwei Zheng et al.
Environment international, 42, 105-116 (2011-05-24)
Pentachlorophenol (PCP) has been widely used for killing snails in areas of China where schistosomiasis is epidemic. With the re-emergence of schistosomiasis, the warranted production and consumption of PCP has inevitably resulted in persistent environmental contamination by it and its
Satyendra Kumar Garg et al.
Environmental science and pollution research international, 20(4), 2288-2304 (2012-08-07)
It is the first report in which a novel psychrotrophic Pseudomonas putida SKG-1 strain was evaluated for simultaneous bioremediation of pentachlorophenol and Cr(6+) under various cultural and nutritional conditions. Pentachlorophenol (PCP) dechlorination products, bacterial structure, and functional groups were characterized
Min Yang et al.
Journal of hazardous materials, 237-238, 63-70 (2012-09-12)
In this study, an imprinted silica matrix of pentachlorophenol (PCP) co-loaded with Fe(3)O(4) nanoparticles and ZnS:Mn(2+) quantum dots (QDs) was fabricated. The introduction of Fe(3)O(4) nanoparticles to the imprinted matrix provided an easy way to separate PCP under an external

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service