Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N12707

2,4′-DDE

analytical standard

Synonym(s):

2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethene, o,p′-DDE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8Cl4
CAS Number:
Molecular Weight:
318.03
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 50 mg

manufacturer/tradename

Chem Service, Inc. N-12707-50MG

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

InChI

1S/C14H8Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8H

InChI key

ZDYJWDIWLRZXDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Benecke et al.
European journal of clinical pharmacology, 41(3), 259-261 (1991-01-01)
Mitotane (o,p'-DDD) can be used for the treatment of various adrenocortical diseases such as Cushing's syndrome, but the usual doses of 6-8 g per day are often associated with severe adverse effects. This paper reports the results of much lower
D B Carlson et al.
Environmental health perspectives, 108(3), 249-255 (2000-03-08)
Fish sexual development is sensitive to exogenous hormone manipulation, and salmonids have been used extensively as environmental sentinels and models for biomedical research. We simulated maternal transfer of contaminants by microinjecting rainbow trout (Oncorhynchus mykiss) and chinook salmon (Oncorhynchus tshawytscha)
P Balaguer et al.
The Science of the total environment, 233(1-3), 47-56 (1999-09-24)
In order to characterize the estrogenic activity of chemicals, we established complementary in vitro recombinant receptor-reporter gene assays in stably transfected MCF-7 and HeLa cells. MCF-7 cells which express the endogenous estrogen receptor alpha (ER alpha) were stably transfected with
W Foster et al.
The Journal of clinical endocrinology and metabolism, 85(8), 2954-2957 (2000-08-18)
Man-made chemicals that have been shown to modulate endocrine function in animal models, so-called "endocrine disrupters", are suspected to play a role in the development of male reproductive tract abnormalities and neurobehaviroal deficits in children. However in utero exposure to
Diana M Papoulias et al.
Environmental health perspectives, 111(1), 29-32 (2003-01-08)
Despite being banned in many countries, dichlorodiphenyltrichloroethane (DDT) and its metabolites dichlorodiphenyldichloroethylene (DDE) and dichlorodiphenyldichloroethane (DDD) continue to be found in fish tissues at concentrations of concern. Like o,p -DDT, o,p -DDE is estrogenic and is believed to exert its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service