Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N12019

Fosetyl-aluminum

technical grade

Synonym(s):

Aluminum tris(O-ethylphosphonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HP(O)(OCH2CH3)O]3Al
CAS Number:
Molecular Weight:
354.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

technical grade

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. N-12019-100MG

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[Al+3].CCO[PH]([O-])=O.CCO[PH]([O-])=O.CCO[PH]([O-])=O

InChI

1S/3C2H7O3P.Al/c3*1-2-5-6(3)4;/h3*6H,2H2,1H3,(H,3,4);/q;;;+3/p-3

InChI key

ZKZMJOFIHHZSRW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Other Notes

Does not come with a purity value or COA.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Quest et al.
Regulatory toxicology and pharmacology : RTP, 14(1), 3-11 (1991-08-01)
Aliette, a fungicide compound, was evaluated for carcinogenic potential by the Health Effects Division of the Office of Pesticide Programs using a consensus peer review process and EPA's guidelines for risk assessment. Aliette was categorized as a group C (possible
Louise R Cooke et al.
Pest management science, 58(1), 17-25 (2002-02-13)
Foliar sprays of potato plants with phosphonic acid (partially neutralised with potassium hydroxide to pH 6.4) substantially reduced infection of the tubers by Phytophthora infestans, the cause of late blight, in glasshouse and field experiments over a 4-year period. Healthy
Occupational exposure to fosetyl-Al fungicide during spraying of ornamentals in greenhouses.
R A Fenske et al.
Archives of environmental contamination and toxicology, 16(5), 615-621 (1987-09-01)
T Deckers et al.
Communications in agricultural and applied biological sciences, 75(4), 569-576 (2010-01-01)
Fire blight, caused by the bacterium Erwinia amylovora (Burill Winslow et al.), is a very important bacterial disease on apple and pear orchards with devastating effects in some production area and in some years. Fire blight control consists in a
Félix Hernández et al.
Journal of AOAC International, 86(4), 832-838 (2003-09-26)
This paper describes a new method for the sensitive and selective determination of fosetyl-aluminum (Al) residues in vegetable samples. The method involves extraction with water by using a high-speed blender and subsequent injection of the 5-fold diluted extract into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service